Tisk…

3,6-Dibromo-9-heptyl-9H-carbazole 98%

BLDPBD00830146-100EA 855 CZK
BLDPBD00830146-100 BLDPBD00830146-250 BLDPBD00830146-1G
3,6-Dibromo-9-heptyl-9H-carbazole 98%
3,6-Dibromo-9-heptyl-9H-carbazole

Storage condition: Sealed in dry, Room Temperature

Vzorec: C₁₉H₂₁Br₂N
MW: 423,19 g/mol
Storage Temperature: Ambient
MDL Number: MFCD31618132
CAS číslo: 1103535-99-4

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR