Tisk…

Methyl 4-carbamoyl-3-nitrobenzoate 97%

BLDPBD00830190-250EA 3650 CZK
BLDPBD00830190-250 BLDPBD00830190-100
Methyl 4-carbamoyl-3-nitrobenzoate 97%
Methyl 4-carbamoyl-3-nitrobenzoate

Storage condition: Sealed in dry, Room Temperature

Vzorec: C₉H₈N₂O₅
MW: 224,1702 g/mol
Storage Temperature: Ambient
MDL Number: MFCD31653743
CAS číslo: 138569-08-1

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR