Tisk…

(S)-1-(5-Chloro-2-fluorophenyl)ethanamine hydrochloride 98%

BLDPBD00830944-100EA 4390 CZK
BLDPBD00830944-100 BLDPBD00830944-250
(S)-1-(5-Chloro-2-fluorophenyl)ethanamine hydrochloride 98%
(S)-1-(5-Chloro-2-fluorophenyl)ethanamine hydrochloride

Storage condition: Inert atmosphere, Room Temperature

Vzorec: C₈H₁₀Cl₂FN
MW: 210,0761 g/mol
Storage Temperature: Ambient
MDL Number: MFCD22741298
CAS číslo: 1423040-76-9

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR