Tisk…

tert-Butyl 14-bromo-3,6,9,12-tetraoxatetradecanoate 95%

BLDPBD00832033-5GEA 22410 CZK
BLDPBD00832033-5G BLDPBD00832033-1G BLDPBD00832033-250
tert-Butyl 14-bromo-3,6,9,12-tetraoxatetradecanoate 95%
tert-Butyl 14-bromo-3,6,9,12-tetraoxatetradecanoate
Vzorec: C₁₄H₂₇BrO₆
MW: 371,26 g/mol
MDL Number: MFCD27977496
CAS číslo: 1807505-29-8

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR