Tisk…

DIsodium phenyl phosphate hydrate 98%

BLDPBD00834275-5GEA 599 CZK
BLDPBD00834275-5G
DIsodium phenyl phosphate hydrate 98%
DIsodium phenyl phosphate hydrate

Storage condition: Inert atmosphere, Room Temperature

Vzorec: C₆H₇Na₂O₅P
MW: 236,0701 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00149081
CAS číslo: 1260375-37-8

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR