Tisk…

3-Chloro-5-fluoro-2-methylbenzoic acid 95%

BLDPBD00834398-5GEA 2800 CZK
BLDPBD00834398-5G BLDPBD00834398-100 BLDPBD00834398-1G BLDPBD00834398-25G
3-Chloro-5-fluoro-2-methylbenzoic acid 95%
3-Chloro-5-fluoro-2-methylbenzoic acid

Storage condition: Sealed in dry, Room Temperature

Vzorec: C₈H₆ClFO₂
MW: 188,5835 g/mol
Storage Temperature: Ambient
MDL Number: MFCD19440350
CAS číslo: 1379210-16-8

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR