Tisk…

N,N-Dipropylethylamine 98%

BLDPBD00837284-1GEA 456 CZK
BLDPBD00837284-1G BLDPBD00837284-25G BLDPBD00837284-5G
N,N-Dipropylethylamine 98%
N,N-Dipropylethylamine

Storage condition: Inert atmosphere, Room Temperature

Vzorec: C₈H₁₉N
MW: 129,25 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00048685
CAS číslo: 20634-92-8

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR