Tisk…

(S)-1-Amino-2,3-dihydro-1H-inden-5-ol hydrochloride 95%

BLDPBD00838976-100EA 9440 CZK
BLDPBD00838976-100 BLDPBD00838976-250 BLDPBD00838976-50M
(S)-1-Amino-2,3-dihydro-1H-inden-5-ol hydrochloride 95%
(S)-1-Amino-2,3-dihydro-1H-inden-5-ol hydrochloride

Storage condition: Inert atmosphere, Room Temperature

Vzorec: C₉H₁₂ClNO
MW: 185,6507 g/mol
Storage Temperature: Ambient
MDL Number: MFCD26585387
CAS číslo: 2703745-66-6

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR