Tisk…

(S)-1-(2,6-dimethylphenyl)ethan-1-amine 95%

BLDPBD00839972-250EA 9320 CZK
BLDPBD00839972-250 BLDPBD00839972-50M BLDPBD00839972-1G BLDPBD00839972-100
(S)-1-(2,6-dimethylphenyl)ethan-1-amine 95%
(S)-1-(2,6-dimethylphenyl)ethan-1-amine

Storage condition: Keep in dark place, Inert atmosphere, Room temperature

Vzorec: C₁₀H₁₅N
MW: 149,2328 g/mol
Storage Temperature: Ambient
MDL Number: MFCD08057435
CAS číslo: 1213479-78-7

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR