Tisk…

(S)-N-Methyl-1-(m-tolyl)ethanamine hydrochloride 95%

BLDPBD00842235-250EA 11520 CZK
BLDPBD00842235-250 BLDPBD00842235-1G BLDPBD00842235-100
(S)-N-Methyl-1-(m-tolyl)ethanamine hydrochloride 95%
(S)-N-Methyl-1-(m-tolyl)ethanamine hydrochloride

Storage condition: Inert atmosphere, 2-8°C

Vzorec: C₁₀H₁₆ClN
MW: 185,7000 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD31706567
CAS číslo: 2241594-13-6

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR