Tisk…

N-(3,3-Difluorocyclobutyl)formamide 98%

BLDPBD00842239-100EA 1600 CZK
BLDPBD00842239-100 BLDPBD00842239-250 BLDPBD00842239-1G
N-(3,3-Difluorocyclobutyl)formamide 98%
N-(3,3-Difluorocyclobutyl)formamide

Storage condition: Sealed in dry, 2-8°C

Vzorec: C₅H₇F₂NO
MW: 135,112 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD24619684
CAS číslo: 1355328-30-1

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR