Tisk…

1-(4-Fluorophenyl)but-3-enylamine hcl 95%

BLDPBD00842470-100EA 4940 CZK
BLDPBD00842470-100 BLDPBD00842470-250
1-(4-Fluorophenyl)but-3-enylamine hcl 95%
1-(4-Fluorophenyl)but-3-enylamine hcl

Storage condition: Inert atmosphere, Room Temperature

Vzorec: C₁₀H₁₃ClFN
MW: 201,6683 g/mol
Storage Temperature: Ambient
MDL Number: MFCD12068564
CAS číslo: 1159825-73-6

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR