Tisk…

2,3,4,9-Tetrahydro-1H-pyrido[3,4-b]indole-1,3-dicarboxylic acid 95%

BLDPBD00842969-1GEA 5360 CZK
BLDPBD00842969-1G BLDPBD00842969-250 BLDPBD00842969-100
2,3,4,9-Tetrahydro-1H-pyrido[3,4-b]indole-1,3-dicarboxylic acid 95%
2,3,4,9-Tetrahydro-1H-pyrido[3,4-b]indole-1,3-dicarboxylic acid

Storage condition: Keep in dark place, Sealed in dry, Store in freezer, under -20°C

Vzorec: C₁₃H₁₂N₂O₄
MW: 260,2454 g/mol
Storage Temperature: Freezer
MDL Number: MFCD00603182
CAS číslo: 59132-30-8

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR