Tisk…

Nalidixic acid sodium salt 98%

BLDPBD00843646-25GEA 3230 CZK
BLDPBD00843646-25G BLDPBD00843646-1G BLDPBD00843646-5G
Nalidixic acid sodium salt 98%
Nalidixic acid sodium salt

Storage condition: Inert atmosphere, Room Temperature

Vzorec: C₁₂H₁₁N₂NaO₃
MW: 254,22 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00064376
CAS číslo: 3374-05-8

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR