Tisk…

6-Bromo-3-fluoroimidazo[1,2-a]pyridine 98%

BLDPBD00843937-100EA 2110 CZK
BLDPBD00843937-100 BLDPBD00843937-1G BLDPBD00843937-250 BLDPBD00843937-5G
6-Bromo-3-fluoroimidazo[1,2-a]pyridine 98%
6-Bromo-3-fluoroimidazo[1,2-a]pyridine

Storage condition: Inert atmosphere, 2-8°C

Vzorec: C₇H₄BrFN₂
MW: 215,0225 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD30829494
CAS číslo: 2001563-56-8

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR