Tisk…

3-Cyclopropyl-4-methyl-1H-pyrazole-5-carboxylic acid 95%

BLDPBD00844726-50MEA 1970 CZK
BLDPBD00844726-50M BLDPBD00844726-1G BLDPBD00844726-250 BLDPBD00844726-100
3-Cyclopropyl-4-methyl-1H-pyrazole-5-carboxylic acid 95%
3-Cyclopropyl-4-methyl-1H-pyrazole-5-carboxylic acid

Storage condition: Sealed in dry, 2-8°C

Vzorec: C₈H₁₀N₂O₂
MW: 166,1700 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD20535329
CAS číslo: 1867157-28-5

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR