Tisk…

(S)-2-Amino-2-(4-bromo-2-methylphenyl)ethan-1-ol hydrochloride 98+%

BLDPBD00847274-100EA 6760 CZK
BLDPBD00847274-100 BLDPBD00847274-250
(S)-2-Amino-2-(4-bromo-2-methylphenyl)ethan-1-ol hydrochloride 98+%
(S)-2-Amino-2-(4-bromo-2-methylphenyl)ethan-1-ol hydrochloride

Storage condition: Inert atmosphere, Room Temperature

Vzorec: C₉H₁₃BrClNO
MW: 266,5626 g/mol
Storage Temperature: Ambient
MDL Number: MFCD31618089
CAS číslo: 2250243-40-2

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR