Tisk…

8-(Trifluoromethoxy)quinoline-5-carboxylic acid 95%

BLDPBD00847962-100EA 10320 CZK
BLDPBD00847962-100 BLDPBD00847962-1G BLDPBD00847962-250
8-(Trifluoromethoxy)quinoline-5-carboxylic acid 95%
8-(Trifluoromethoxy)quinoline-5-carboxylic acid

Storage condition: Sealed in dry, Room Temperature

Vzorec: C₁₁H₆F₃NO₃
MW: 257,1655 g/mol
Storage Temperature: Ambient
MDL Number: MFCD31729491
CAS číslo: 2241594-63-6

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR