Tisk…

tert-Butyl (2-(2-(2-(2-bromoacetamido)ethoxy)ethoxy)ethyl)carbamate 95%

BLDPBD00848385-250EA 1460 CZK
BLDPBD00848385-250 BLDPBD00848385-1G BLDPBD00848385-5G BLDPBD00848385-100
tert-Butyl (2-(2-(2-(2-bromoacetamido)ethoxy)ethoxy)ethyl)carbamate 95%
tert-Butyl (2-(2-(2-(2-bromoacetamido)ethoxy)ethoxy)ethyl)carbamate

Storage condition: Inert atmosphere, 2-8°C

Vzorec: C₁₃H₂₅BrN₂O₅
MW: 369,252 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD28142467
CAS číslo: 182244-33-3

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR