Tisk…

(R)-Methyl 2-amino-4-((tert-butoxycarbonyl)amino)butanoate hydrochloride 95%

BLDPBD00849350-250EA 1290 CZK
BLDPBD00849350-250 BLDPBD00849350-1G BLDPBD00849350-5G
(R)-Methyl 2-amino-4-((tert-butoxycarbonyl)amino)butanoate hydrochloride 95%
(R)-Methyl 2-amino-4-((tert-butoxycarbonyl)amino)butanoate hydrochloride

Storage condition: Inert atmosphere, 2-8°C

Vzorec: C₁₀H₂₁ClN₂O₄
MW: 268,7377 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD08275859
CAS číslo: 1052649-77-0

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR