Tisk…

19-Bromo-2,5,8,11,14,17-hexaoxanonadecane 95%

BLDPBD00849446-5GEA 13370 CZK
BLDPBD00849446-5G BLDPBD00849446-1G BLDPBD00849446-250
19-Bromo-2,5,8,11,14,17-hexaoxanonadecane 95%
19-Bromo-2,5,8,11,14,17-hexaoxanonadecane

Storage condition: Sealed in dry, 2-8°C

Vzorec: C₁₃H₂₇BrO₆
MW: 359,2539 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD22683286
CAS číslo: 125562-29-0

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR