Tisk…

2-(2-(3-(tert-Butoxy)-3-oxopropoxy)ethoxy)ethyl acrylate 95%

BLDPBD00851441-250EA 2940 CZK
BLDPBD00851441-250 BLDPBD00851441-1G BLDPBD00851441-100 BLDPBD00851441-5G
2-(2-(3-(tert-Butoxy)-3-oxopropoxy)ethoxy)ethyl acrylate 95%
2-(2-(3-(tert-Butoxy)-3-oxopropoxy)ethoxy)ethyl acrylate

Storage condition: Sealed in dry, Store in freezer, under -20°C

Vzorec: C₁₄H₂₄O₆
MW: 288,3368 g/mol
Storage Temperature: Freezer
MDL Number: MFCD28015773
CAS číslo: 1807518-62-2

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR