Tisk…

n-Hexadecyl-β-D-maltoside 98%

BLDPBD00852473-1GEA 5390 CZK
BLDPBD00852473-1G BLDPBD00852473-100 BLDPBD00852473-250
n-Hexadecyl-β-D-maltoside 98%
n-Hexadecyl-β-D-maltoside

Storage condition: Inert atmosphere, 2-8°C

Vzorec: C₂₈H₅₄O₁₁
MW: 566,72 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD01310880
CAS číslo: 98064-96-1

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR