Tisk…

Ethyl 2-bromo-2-(3-(trifluoromethyl)phenyl)acetate 95%

BLDPBD00853405-250EA 8470 CZK
BLDPBD00853405-250 BLDPBD00853405-100
Ethyl 2-bromo-2-(3-(trifluoromethyl)phenyl)acetate 95%
Ethyl 2-bromo-2-(3-(trifluoromethyl)phenyl)acetate

Storage condition: Inert atmosphere, 2-8°C

Vzorec: C₁₁H₁₀BrF₃O₂
MW: 311,0951 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD27950529
CAS číslo: 41023-25-0

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR