Tisk…

1-(tert-Butoxycarbonyl)-5'-methyl-4',5'-dihydrospiro[piperidine-4,7'-thieno[2,3-c]pyran]-2'-carboxylic acid 95%

BLDPBD00853995-25MEA 3480 CZK
BLDPBD00853995-25M BLDPBD00853995-250 BLDPBD00853995-50M BLDPBD00853995-5G BLDPBD00853995-100 BLDPBD00853995-1G
1-(tert-Butoxycarbonyl)-5'-methyl-4',5'-dihydrospiro[piperidine-4,7'-thieno[2,3-c]pyran]-2'-carboxylic acid 95%
1-(tert-Butoxycarbonyl)-5'-methyl-4',5'-dihydrospiro[piperidine-4,7'-thieno[2,3-c]pyran]-2'-carboxylic acid

Storage condition: Sealed in dry, 2-8°C

Vzorec: C₁₈H₂₅NO₅S
MW: 367,4598 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD31555888
CAS číslo: 2088333-65-5

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR