Tisk…

6,8-Difluorothiochroman-4-one 95%

BLDPBD00854165-100EA 3770 CZK
BLDPBD00854165-100 BLDPBD00854165-250 BLDPBD00854165-1G
6,8-Difluorothiochroman-4-one 95%
6,8-Difluorothiochroman-4-one

Storage condition: Sealed in dry, Room Temperature

Vzorec: C₉H₆F₂OS
MW: 200,2052 g/mol
Storage Temperature: Ambient
MDL Number: MFCD11642634
CAS číslo: 1097779-72-0

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR