Tisk…

5-Chloroquinoline-3-carbonitrile 98%

BLDPBD00854707-250EA 14340 CZK
BLDPBD00854707-250 BLDPBD00854707-1G BLDPBD00854707-100
5-Chloroquinoline-3-carbonitrile 98%
5-Chloroquinoline-3-carbonitrile

Storage condition: Sealed in dry, Room Temperature

Vzorec: C₁₀H₅ClN₂
MW: 188,6131 g/mol
Storage Temperature: Ambient
MDL Number: MFCD31556078
CAS číslo: 2102411-05-0

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR