Tisk…

(S)-1-(4-Bromo-2,6-difluorophenyl)ethanamine 95%

BLDPBD00854789-50MEA 3600 CZK
BLDPBD00854789-50M
(S)-1-(4-Bromo-2,6-difluorophenyl)ethanamine 95%
(S)-1-(4-Bromo-2,6-difluorophenyl)ethanamine

Storage condition: Keep in dark place, Inert atmosphere, 2-8°C

Vzorec: C₈H₈BrF₂N
MW: 236,0566 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD07772571
CAS číslo: 1241677-54-2

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR