Tisk…

Tributyl(2-methoxyethyl)phosphonium bis(trifluoromethanesulphonyl)imide 98%

BLDPBD00854827-5GEA 2110 CZK
BLDPBD00854827-5G BLDPBD00854827-1G BLDPBD00854827-25G
Tributyl(2-methoxyethyl)phosphonium bis(trifluoromethanesulphonyl)imide 98%
Tributyl(2-methoxyethyl)phosphonium bis(trifluoromethanesulphonyl)imide

Storage condition: Inert atmosphere, Room Temperature

Vzorec: C₁₇H₃₄F₆NO₅PS₂
MW: 541,55 g/mol
Storage Temperature: Ambient
MDL Number: MFCD11112190
CAS číslo: 959698-44-3

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR