Tisk…

2-((2,6-Dimethylphenyl)amino)-N,N-diethyl-2-oxoethan-1-amine oxide 98%

BLDPBD00857740-100EA 6130 CZK
BLDPBD00857740-100 BLDPBD00857740-1G BLDPBD00857740-250
2-((2,6-Dimethylphenyl)amino)-N,N-diethyl-2-oxoethan-1-amine oxide 98%
2-((2,6-Dimethylphenyl)amino)-N,N-diethyl-2-oxoethan-1-amine oxide

Storage condition: Sealed in dry, 2-8°C

Vzorec: C₁₄H₂₂N₂O₂
MW: 250,3367 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD22125322
CAS číslo: 2903-45-9

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR