Tisk…

6-Bromo-5-fluoro-2,3-dihydro-1H-inden-1-ol 95%

BLDPBD00858495-250EA 9440 CZK
BLDPBD00858495-250 BLDPBD00858495-1G BLDPBD00858495-100
6-Bromo-5-fluoro-2,3-dihydro-1H-inden-1-ol 95%
6-Bromo-5-fluoro-2,3-dihydro-1H-inden-1-ol

Storage condition: Sealed in dry, Room Temperature

Vzorec: C₉H₈BrFO
MW: 231,0616 g/mol
Storage Temperature: Ambient
MDL Number: MFCD30829615
CAS číslo: 2089649-48-7

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR