Tisk…

(S)-1-(2-Chloro-4-methoxyphenyl)ethan-1-amine hydrochloride 95%

BLDPBD00859707-250EA 10380 CZK
BLDPBD00859707-250 BLDPBD00859707-100
(S)-1-(2-Chloro-4-methoxyphenyl)ethan-1-amine hydrochloride 95%
(S)-1-(2-Chloro-4-methoxyphenyl)ethan-1-amine hydrochloride

Storage condition: Inert atmosphere, Room Temperature

Vzorec: C₉H₁₃Cl₂NO
MW: 222,1116 g/mol
Storage Temperature: Ambient
MDL Number: MFCD31706462
CAS číslo: 2241594-14-7

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR