Tisk…

(S)-1-Benzyl 4-methyl 2-((tert-butoxycarbonyl)amino)succinate 95%

BLDPBD00859930-1GEA 3400 CZK
BLDPBD00859930-1G BLDPBD00859930-250 BLDPBD00859930-100
(S)-1-Benzyl 4-methyl 2-((tert-butoxycarbonyl)amino)succinate 95%
(S)-1-Benzyl 4-methyl 2-((tert-butoxycarbonyl)amino)succinate

Storage condition: Sealed in dry, 2-8°C

Vzorec: C₁₇H₂₃NO₆
MW: 337,3676 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD27578330
CAS číslo: 158201-15-1

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR