Tisk…

2-Bromo-5H,6H,7H-pyrrolo[1,2-a]imidazole 98%

BLDPBD00862423-250EA 4560 CZK
BLDPBD00862423-250 BLDPBD00862423-5G BLDPBD00862423-100 BLDPBD00862423-1G
2-Bromo-5H,6H,7H-pyrrolo[1,2-a]imidazole 98%
2-Bromo-5H,6H,7H-pyrrolo[1,2-a]imidazole

Storage condition: Inert atmosphere, 2-8°C

Vzorec: C₆H₇BrN₂
MW: 187,0372 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD29047774
CAS číslo: 1525619-20-8

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR