Tisk…

Potassium 2-fluorobenzyl-trifluoroborate 97%

BLDPBD00863361-100EA 2460 CZK
BLDPBD00863361-100 BLDPBD00863361-250
Potassium 2-fluorobenzyl-trifluoroborate 97%
Potassium 2-fluorobenzyl-trifluoroborate

Storage condition: Inert atmosphere, Room Temperature

Vzorec: C₇H₆BF₄·K
MW: 216,025 g/mol
Storage Temperature: Ambient
MDL Number: MFCD09993122
CAS číslo: 1246073-48-2

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR