Tisk…

Methyl 4-iodo-1,2-thiazole-3-carboxylate 95%

BLDPBD00866094-1GEA 28820 CZK
BLDPBD00866094-1G BLDPBD00866094-100 BLDPBD00866094-250
Methyl 4-iodo-1,2-thiazole-3-carboxylate 95%
Methyl 4-iodo-1,2-thiazole-3-carboxylate

Storage condition: Keep in dark place, Sealed in dry, 2-8°C

Vzorec: C₅H₄INO₂S
MW: 269,0602 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD29060514
CAS číslo: 397329-61-2

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR