Tisk…

(1S)-1-(4-Phenylphenyl)ethan-1-amine hydrochloride 95%

BLDPBD00867223-100EA 5880 CZK
BLDPBD00867223-100 BLDPBD00867223-1G BLDPBD00867223-250
(1S)-1-(4-Phenylphenyl)ethan-1-amine hydrochloride 95%
(1S)-1-(4-Phenylphenyl)ethan-1-amine hydrochloride

Storage condition: Inert atmosphere, Room Temperature

Vzorec: C₁₄H₁₆ClN
MW: 233,7365 g/mol
Storage Temperature: Ambient
MDL Number: MFCD20233489
CAS číslo: 1354970-76-5

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR