Tisk…

(3aS,4S,6aR)-4-(5-Hydroxypentyl)tetrahydro-1H-thieno[3,4-d]imidazol-2(3H)-one 96%

BLDPBD00868592-250EA 7500 CZK
BLDPBD00868592-250 BLDPBD00868592-100
(3aS,4S,6aR)-4-(5-Hydroxypentyl)tetrahydro-1H-thieno[3,4-d]imidazol-2(3H)-one 96%
(3aS,4S,6aR)-4-(5-Hydroxypentyl)tetrahydro-1H-thieno[3,4-d]imidazol-2(3H)-one

Storage condition: Keep in dark place, Inert atmosphere, Store in freezer, under -20°C

Vzorec: C₁₀H₁₈N₂O₂S
MW: 230,3271 g/mol
Storage Temperature: Freezer
MDL Number: MFCD30532025
CAS číslo: 53906-36-8

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR