Tisk…

2-(Cyanosulphanyl)-1-(3,4-dihydroxyphenyl)ethan-1-one 95%

BLDPBD00868745-100EA 2880 CZK
BLDPBD00868745-100 BLDPBD00868745-250 BLDPBD00868745-1G
2-(Cyanosulphanyl)-1-(3,4-dihydroxyphenyl)ethan-1-one 95%
2-(Cyanosulphanyl)-1-(3,4-dihydroxyphenyl)ethan-1-one

Storage condition: Sealed in dry, 2-8°C

Vzorec: C₉H₇NO₃S
MW: 209,2218 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD22415334
CAS číslo: 101714-41-4

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR