Tisk…

2-(2,4-Difluorophenyl)-2,2-difluoroethan-1-amine hydrochloride 95%

BLDPBD00872529-1GEA 15080 CZK
BLDPBD00872529-1G BLDPBD00872529-250 BLDPBD00872529-50M BLDPBD00872529-100
2-(2,4-Difluorophenyl)-2,2-difluoroethan-1-amine hydrochloride 95%
2-(2,4-Difluorophenyl)-2,2-difluoroethan-1-amine hydrochloride

Storage condition: Inert atmosphere, Room Temperature

Vzorec: C₈H₈ClF₄N
MW: 229,6024 g/mol
Storage Temperature: Ambient
MDL Number: MFCD30502238
CAS číslo: 2060062-92-0

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR