Tisk…

1-Bromo-4-(butane-1-sulphinyl)benzene 95%

BLDPBD00873503-1GEA 684 CZK
BLDPBD00873503-1G
1-Bromo-4-(butane-1-sulphinyl)benzene 95%
1-Bromo-4-(butane-1-sulphinyl)benzene

Storage condition: Keep in dark place, Inert atmosphere, Room temperature

Vzorec: C₁₀H₁₃BrOS
MW: 261,1786 g/mol
Storage Temperature: Ambient
MDL Number: MFCD29043834
CAS číslo: 1865653-34-4

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR