Tisk…

3-Bromo-2-(tert-butoxy)pyridine 95%

BLDPBD00879465-100EA 2340 CZK
BLDPBD00879465-100 BLDPBD00879465-250 BLDPBD00879465-1G
3-Bromo-2-(tert-butoxy)pyridine 95%
3-Bromo-2-(tert-butoxy)pyridine

Storage condition: Sealed in dry, Room Temperature

Vzorec: C₉H₁₂BrNO
MW: 230,1017 g/mol
Storage Temperature: Ambient
MDL Number: MFCD14651779
CAS číslo: 1245898-81-0

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR