Tisk…

5-Chloro-N,2-dimethylaniline 95%

BLDPBD00880175-250EA 1690 CZK
BLDPBD00880175-250 BLDPBD00880175-5G BLDPBD00880175-1G
5-Chloro-N,2-dimethylaniline 95%
5-Chloro-N,2-dimethylaniline

Storage condition: Keep in dark place, Inert atmosphere, Room temperature

Vzorec: C₈H₁₀ClN
MW: 155,6247 g/mol
Storage Temperature: Ambient
MDL Number: MFCD03030402
CAS číslo: 41456-54-6

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR