Tisk…

(6-Acetylpyridin-3-yl)boronic acid 95%

BLDPBD00880217-1GEA 5020 CZK
BLDPBD00880217-1G BLDPBD00880217-250 BLDPBD00880217-5G BLDPBD00880217-100
(6-Acetylpyridin-3-yl)boronic acid 95%
(6-Acetylpyridin-3-yl)boronic acid

Storage condition: Inert atmosphere, 2-8°C

Vzorec: C₇H₈BNO₃
MW: 164,9543 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD07374894
CAS číslo: 2682066-26-6

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR