Tisk…

(R)-1-(4-iodophenyl)ethan-1-ol 95%

BLDPBD00880671-250EA 8100 CZK
BLDPBD00880671-250 BLDPBD00880671-100 BLDPBD00880671-1G
(R)-1-(4-iodophenyl)ethan-1-ol 95%
(R)-1-(4-iodophenyl)ethan-1-ol

Storage condition: Keep in dark place, Sealed in dry, 2-8°C

Vzorec: C₈H₉IO
MW: 248,0609 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD09863732
CAS číslo: 220089-24-7

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR