Tisk…

5-Ethyl-6-methyl-2-thioxo-2,3-dihydropyrimidin-4(1H)-one 95%

BLDPBD00881292-1GEA 9670 CZK
BLDPBD00881292-1G BLDPBD00881292-250 BLDPBD00881292-100
5-Ethyl-6-methyl-2-thioxo-2,3-dihydropyrimidin-4(1H)-one 95%
5-Ethyl-6-methyl-2-thioxo-2,3-dihydropyrimidin-4(1H)-one

Storage condition: Sealed in dry, Room Temperature

Vzorec: C₇H₁₀N₂OS
MW: 170,2321 g/mol
Storage Temperature: Ambient
MDL Number: MFCD01677038
CAS číslo: 39083-15-3

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR