Tisk…

1-Benzoyl-4-(chloroacetyl)piperazine 95%

BLDPBD00881621-1GEA 5700 CZK
BLDPBD00881621-1G BLDPBD00881621-5G BLDPBD00881621-250
1-Benzoyl-4-(chloroacetyl)piperazine 95%
1-Benzoyl-4-(chloroacetyl)piperazine

Storage condition: Inert atmosphere, 2-8°C

Vzorec: C₁₃H₁₅ClN₂O₂
MW: 266,7234 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD04116666
CAS číslo: 69463-43-0

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR