Tisk…

tert-Butyl N-(4-bromo-3-fluorophenyl)-N-methylcarbamate 95%

BLDPBD00881847-100EA 2230 CZK
BLDPBD00881847-100 BLDPBD00881847-250 BLDPBD00881847-1G
tert-Butyl N-(4-bromo-3-fluorophenyl)-N-methylcarbamate 95%
tert-Butyl N-(4-bromo-3-fluorophenyl)-N-methylcarbamate

Storage condition: Sealed in dry, 2-8°C

Vzorec: C₁₂H₁₅BrFNO₂
MW: 304,1554 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD29043959
CAS číslo: 1881330-84-2

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR