Tisk…

(R)-2-(2,5-Dichlorophenyl)pyrrolidine hydrochloride 98%

BLDPBD00882133-250EA 10950 CZK
BLDPBD00882133-250 BLDPBD00882133-1G BLDPBD00882133-100
(R)-2-(2,5-Dichlorophenyl)pyrrolidine hydrochloride 98%
(R)-2-(2,5-Dichlorophenyl)pyrrolidine hydrochloride

Storage condition: Inert atmosphere, Room Temperature

Vzorec: C₁₀H₁₁Cl₂N·ClH
MW: 252,568 g/mol
Storage Temperature: Ambient
MDL Number: MFCD26583830
CAS číslo: 2241594-21-6

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR