Tisk…

2-(2-((tert-Butoxycarbonyl)amino)ethoxy)ethyl methanesulphonate 95%

BLDPBD00892247-250EA 713 CZK
BLDPBD00892247-250 BLDPBD00892247-1G
2-(2-((tert-Butoxycarbonyl)amino)ethoxy)ethyl methanesulphonate 95%
2-(2-((tert-Butoxycarbonyl)amino)ethoxy)ethyl methanesulphonate

Storage condition: Sealed in dry, 2-8°C

Vzorec: C₁₀H₂₁NO₆S
MW: 283,3418 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD24465573
CAS číslo: 302331-20-0

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR